siRNA Details
Gene Symbol: Scgb1b27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009596.1 | NP_033726.1 | 2 | 147 | ||
GenBank mRNA | AF039064.1 | - | 1 | 80 | ||
BC016132.1 | - | 2 | 147 | 4922791 | ||
BC132436.1 | - | 2 | 231 | 40130713 | ||
GU269231.1 | - | 2 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
