siRNA Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009610.2 | NP_033740.2 | 3 | 303 | ||
GenBank mRNA | AK146631.1 | - | 3 | 276 | ||
AK168649.1 | - | 3 | 276 | |||
BC002042.1 | - | 3 | 284 | 3591323 | ||
BC049611.1 | - | 3 | 335 | 6773765 | ||
BY778654.1 | - | 3 | 304 | |||
M26689.1 | - | 2 | 232 |

More Information
Other Species Targeted

Gene Ontology Categories:
