siRNA Details
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013456.2 | NP_038484.1 | 5 | 587 | ||
GenBank mRNA | AF093775.1 | - | 5 | 562 | ||
BC111890.1 | - | 5 | 553 | 40060598 |

More Information
Other Species Targeted

Gene Ontology Categories:
