siRNA Details
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277255.1 | NP_001264184.1 | 10 | 1406 | ||
NM_001277257.1 | NP_001264186.1 | 9 | 1345 | |||
NM_001277258.1 | NP_001264187.1 | 9 | 1314 | |||
NM_001277259.1 | NP_001264188.1 | 10 | 1401 | |||
NM_009612.3 | NP_033742.2 | 9 | 1383 | |||
GenBank mRNA | AK134712.1 | - | 7 | 1140 | ||
AK160915.1 | - | 10 | 1374 | |||
AK170870.1 | - | 7 | 1139 | |||
BC014291.1 | - | 8 | 1302 | 3982004 | ||
BC015083.1 | - | 8 | 1264 | 3980151 | ||
CT010330.1 | - | 7 | 1111 | |||
L48015.1 | - | 9 | 1385 | |||
Z31664.1 | - | 9 | 1351 |
