siRNA Details
Gene Symbol: Adam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110778.1 | NP_001104248.1 | 15 | 1459 | ||
NM_009613.2 | NP_033743.2 | 15 | 1459 | |||
GenBank mRNA | AB009676.1 | - | 15 | 1292 | ||
AK157926.1 | - | 15 | 1464 | |||
BC054536.1 | - | 15 | 1319 | 6400084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
