siRNA Details
Gene Symbol: GRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000839.3 | NP_000830.2 | 3 | 772 | ||
XM_011533635.1 | XP_011531937.1 | 2 | 2443 | |||
XM_011533637.1 | XP_011531939.1 | 2 | 768 | |||
XM_011533638.2 | XP_011531940.1 | 3 | 510 | |||
XM_011533639.2 | XP_011531941.1 | 3 | 228 | |||
XM_011533640.2 | XP_011531942.1 | 3 | 181 | |||
XM_011533641.2 | XP_011531943.1 | 3 | 217 | |||
XM_011533642.1 | XP_011531944.1 | 2 | 2443 | |||
XM_017006271.1 | XP_016861760.1 | 3 | 1250 | |||
GenBank mRNA | AB209164.1 | - | 3 | 1262 | ||
AK289984.1 | - | 3 | 765 | |||
AK302671.1 | - | 2 | 531 | |||
AK313056.1 | - | 3 | 749 | |||
AK315816.1 | - | 3 | 767 | |||
AY999299.1 | - | 2 | 569 | |||
BC113615.1 | - | 2 | 556 | 8322667 | ||
BC113619.1 | - | 2 | 556 | 8322671 | ||
BX648951.1 | - | 2 | 2212 | |||
EU432122.1 | - | 2 | 533 | |||
L35318.1 | - | 2 | 535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
