siRNA Details
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009619.4 | NP_033749.2 | 7 | 616 | ||
GenBank mRNA | BC060975.1 | - | 7 | 579 | ||
BC129920.1 | - | 7 | 582 | |||
X64227.1 | - | 7 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
