siRNA Details
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122188.1 | - | 9 | 1005 | ||
AK148174.1 | - | 9 | 908 | |||
AK155672.1 | - | 9 | 915 | |||
BC047156.1 | - | 9 | 900 | 6488707 | ||
U41765.1 | - | 9 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
