siRNA Details
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270996.1 | NP_001257925.1 | 18, 19 | 2203 | ||
NM_007404.2 | NP_031430.2 | 18, 19 | 2203 | |||
GenBank mRNA | AK122188.1 | - | 18, 19 | 2186 | ||
AK148174.1 | - | 18, 19 | 2089 | |||
AK155582.1 | - | 18, 19 | 2166 | |||
AK155672.1 | - | 18, 19 | 2096 | |||
AK161778.1 | - | 18, 19 | 2126 | |||
BC047156.1 | - | 18, 19 | 2081 | 6488707 | ||
BC052710.1 | - | 18, 19 | 2075 | |||
U41765.1 | - | 18, 19 | 2075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
