siRNA Details
Gene Symbol: Gpr182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007412.2 | NP_031438.2 | 2 | 314 | ||
GenBank mRNA | AK031480.1 | - | 249 | |||
BC125279.1 | - | 2 | 185 | |||
BY121440.1 | - | 2 | 315 | |||
D17292.1 | - | 2 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
