siRNA Details
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178803.2 | NP_848918.1 | 7 | 1129 | ||
GenBank mRNA | AB221647.1 | - | 7 | 780 | ||
AF090326.1 | - | 864 | ||||
AK029011.1 | - | 7 | 1131 | |||
AK045838.1 | - | 7 | 895 | |||
AK045987.1 | - | 7 | 895 | |||
AK155272.1 | - | 7 | 861 | |||
BC025467.1 | - | 7 | 1587 | 5351959 | ||
BC031376.1 | - | 8 | 990 | 5356616 | ||
BC057681.1 | - | 7 | 909 | 3499299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
