siRNA Details
Gene Symbol: Crisp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009638.3 | NP_033768.3 | 7, 8 | 658 | ||
GenBank mRNA | AK009449.1 | - | 7, 8 | 659 | ||
BC011150.1 | - | 7, 8 | 641 | 4165956 | ||
L05559.1 | - | 7, 8 | 628 | |||
M92849.1 | - | 7, 8 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
