siRNA Details
Gene Symbol: Afp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007423.4 | NP_031449.3 | 6 | 710 | ||
GenBank mRNA | AK010934.1 | - | 6 | 686 | ||
AK075972.1 | - | 6 | 690 | |||
AK076053.1 | - | 6 | 686 | |||
AK076197.1 | - | 6 | 688 | |||
AK145975.1 | - | 6 | 686 | |||
AK146513.1 | - | 6 | 688 | |||
AK146524.1 | - | 6 | 688 | |||
AK167549.1 | - | 6 | 686 | |||
AK168426.1 | - | 6 | 689 | |||
AK168574.1 | - | 6 | 688 | |||
AK168576.1 | - | 6 | 686 | |||
AK168751.1 | - | 6 | 688 | |||
AK168818.1 | - | 5 | 564 | |||
AK168883.1 | - | 6 | 686 | |||
AK169046.1 | - | 6 | 688 | |||
AK169054.1 | - | 6 | 688 | |||
AK169147.1 | - | 6 | 686 | |||
BC066206.1 | - | 6 | 673 | 30469872 | ||
V00743.1 | - | 6 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
