siRNA Details
Gene Symbol: Agtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007429.5 | NP_031455.1 | 3 | 1027 | ||
GenBank mRNA | AK086334.1 | - | 3 | 1026 | ||
AK140941.1 | - | 3 | 1029 | |||
BC003811.1 | - | 1 | 897 | 3497420 | ||
L32840.1 | - | 3 | 1016 | |||
S67465.1 | - | 3 | 991 | |||
U04828.1 | - | 3 | 991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
