siRNA Details
Gene Symbol: Agtrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301281.1 | NP_001288210.1 | 5 | 1412 | ||
NM_009642.4 | NP_033772.2 | 5 | 1459 | |||
GenBank mRNA | AK036598.1 | - | 5 | 1388 | ||
AK152467.1 | - | 5 | 1459 | |||
BC046820.1 | - | 5 | 1445 | 6400135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
