siRNA Details
Gene Symbol: Alppl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007433.3 | NP_031459.3 | 10 | 1387 | ||
GenBank mRNA | AK145571.1 | - | 11 | 1296 | ||
AY054302.1 | - | 10 | 1228 | |||
BC052874.1 | - | 10 | 1382 | 30033693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
