siRNA Details
Gene Symbol: Alas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291835.1 | NP_001278764.1 | 4 | 1044 | ||
NM_020559.2 | NP_065584.2 | 4 | 657 | |||
GenBank mRNA | AK077152.1 | - | 3 | 348 | ||
AK133283.1 | - | 4 | 1045 | |||
BC022110.1 | - | 4 | 559 | 5133249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
