siRNA Details
Gene Symbol: Alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004002.1 | - | 5 | 495 | ||
AK014153.1 | - | 2 | 136 | |||
AK075967.1 | - | 5 | 478 | |||
BC026209.1 | - | 5 | 482 | 1349819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
