siRNA Details
Gene Symbol: Amd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007444.3 | NP_031470.3 | 1 | 1045 |
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009665.5 | NP_033795.1 | 6, 7 | 1043 | ||
GenBank mRNA | AK146840.1 | - | 6, 7 | 1042 | ||
AK166187.1 | - | 6, 7 | 1037 | |||
AK167423.1 | - | 1, 2 | 93 | |||
AK168165.1 | - | 6, 7 | 1040 | |||
BC011110.1 | - | 6, 7 | 1028 | 3672700 | ||
BC071220.1 | - | 6, 7 | 1026 | 5012910 | ||
BC080791.1 | - | 6, 7 | 807 | 4014056 | ||
BC092072.1 | - | 6, 7 | 1024 | 4505452 | ||
BC138696.1 | - | 6, 7 | 781 | 8861718 | ||
BC138697.1 | - | 6, 7 | 781 | 8861719 | ||
CT010192.1 | - | 6, 7 | 713 | |||
D12780.1 | - | 6, 7 | 1039 | |||
Z14986.1 | - | 6, 7 | 970 |

More Information
Other Species Targeted

Gene Ontology Categories:
