siRNA Details
Gene Symbol: Amd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007444.3 | NP_031470.3 | 1 | 807 |
Gene Symbol: Amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009665.5 | NP_033795.1 | 5, 6 | 805 | ||
GenBank mRNA | AK146840.1 | - | 5, 6 | 804 | ||
AK166187.1 | - | 5, 6 | 799 | |||
AK168165.1 | - | 5, 6 | 802 | |||
AK171238.1 | - | 5, 6 | 804 | |||
BC011110.1 | - | 5, 6 | 790 | 3672700 | ||
BC071220.1 | - | 5, 6 | 788 | 5012910 | ||
BC080791.1 | - | 5, 6 | 569 | 4014056 | ||
BC092072.1 | - | 5, 6 | 786 | 4505452 | ||
BC138696.1 | - | 5, 6 | 543 | 8861718 | ||
BC138697.1 | - | 5, 6 | 543 | 8861719 | ||
CT010192.1 | - | 5, 6 | 475 | |||
D12780.1 | - | 5, 6 | 801 | |||
Z14986.1 | - | 5, 6 | 732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
