siRNA Details
Gene Symbol: Amy2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160150.1 | NP_001153622.1 | 605 |
Gene Symbol: Amy2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160152.1 | NP_001153624.1 | 3 | 605 |
Gene Symbol: Amy2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160151.1 | NP_001153623.1 | 605 |
Gene Symbol: Amy2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042711.2 | NP_001036176.1 | 467 | |||
GenBank mRNA | AK078968.1 | - | 465 | |||
AK133526.1 | - | 468 | ||||
BC094924.1 | - | 455 | 6433956 | |||
BC100579.1 | - | 462 | 6433256 | |||
V00718.1 | - | 468 | ||||
X02576.1 | - | 466 | ||||
X02577.1 | - | 466 | ||||
X02578.1 | - | 466 | ||||
X02579.1 | - | 466 |
Gene Symbol: Amy2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190403.1 | NP_001177332.1 | 3 | 467 | ||
NM_001190404.1 | NP_001177333.1 | 3 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
