siRNA Details
Gene Symbol: Ang | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161731.2 | NP_001155203.1 | 2 | 559 | ||
NM_007447.3 | NP_031473.1 | 2 | 856 | |||
GenBank mRNA | AK159966.1 | - | 2 | 857 | ||
AK171251.1 | - | 2 | 487 | |||
BC055355.1 | - | 2 | 443 | 5136641 | ||
BF721354.1 | - | 2 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
