siRNA Details
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 8 | 1653 | ||
NM_146005.3 | NP_666117.2 | 31 | 4124 | |||
NM_170687.3 | NP_733788.2 | 8 | 1653 | |||
NM_170688.2 | NP_733789.1 | 31 | 4124 | |||
NM_170689.2 | NP_733790.2 | 30 | 4070 | |||
NM_170690.2 | NP_733791.2 | 30 | 4061 | |||
NM_170728.2 | NP_733924.2 | 29 | 4007 | |||
NM_170729.2 | NP_733925.2 | 29 | 4007 | |||
NM_170730.2 | NP_733926.2 | 6 | 1343 | |||
GenBank mRNA | AF212924.1 | - | 6 | 843 | ||
AK035681.1 | - | 8 | 1381 | |||
AK139233.1 | - | 2 | 202 | |||
AK141720.1 | - | 31 | 3979 | |||
AK157735.1 | - | 9 | 1401 | |||
AK162007.1 | - | 8 | 1311 | |||
BC021657.1 | - | 29 | 3877 | 4188590 | ||
BC026152.1 | - | 8 | 1255 | |||
L40631.1 | - | 8 | 1656 | |||
L40632.1 | - | 31 | 3666 | |||
U89274.1 | - | 10 | 1270 | |||
U89275.1 | - | 10 | 1270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
