siRNA Details
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 19 | 3294 | ||
NM_146005.3 | NP_666117.2 | 42 | 6353 | |||
NM_170687.3 | NP_733788.2 | 19 | 3882 | |||
NM_170688.2 | NP_733789.1 | 42 | 5765 | |||
NM_170689.2 | NP_733790.2 | 41 | 6299 | |||
NM_170690.2 | NP_733791.2 | 41 | 6290 | |||
NM_170728.2 | NP_733924.2 | 40 | 5648 | |||
NM_170729.2 | NP_733925.2 | 40 | 6236 | |||
NM_170730.2 | NP_733926.2 | 17 | 2984 | |||
GenBank mRNA | AF212924.1 | - | 17 | 2484 | ||
AK033389.1 | - | 4 | 1180 | |||
AK035681.1 | - | 19 | 3607 | |||
BC021657.1 | - | 40 | 5518 | 4188590 | ||
BC026152.1 | - | 19 | 2896 | |||
BC062122.1 | - | 8 | 3602 | |||
L40631.1 | - | 19 | 3885 | |||
L40632.1 | - | 42 | 5895 | |||
U89274.1 | - | 21 | 2911 | |||
U89275.1 | - | 21 | 3499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
