siRNA Details
Gene Symbol: Slc25a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007450.4 | NP_031476.3 | 2 | 351 | ||
GenBank mRNA | AK051172.1 | - | 2 | 374 | ||
AK078077.1 | - | 2 | 373 | |||
AK203920.1 | - | 1 | 34 | |||
BC003791.1 | - | 2 | 351 | 3599576 | ||
BC026925.1 | - | 2 | 362 | 4500784 | ||
U27315.1 | - | 2 | 350 | |||
X74510.1 | - | 2 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
