siRNA Details
Gene Symbol: Slc25a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002283.1 | - | 2 | 232 | ||
AK003467.1 | - | 2 | 235 | |||
AK012751.1 | - | 2 | 233 | |||
AK017881.1 | - | 2 | 232 | |||
AK088730.1 | - | 2 | 232 | |||
AK188826.1 | - | 2 | 82 | |||
AK195999.1 | - | 2 | 82 | |||
BC004570.1 | - | 2 | 215 | 3586336 | ||
BC086756.1 | - | 2 | 214 | 6818521 | ||
BY280917.1 | - | 2 | 289 | |||
U10404.1 | - | 2 | 155 | |||
U27316.1 | - | 2 | 240 | |||
X70847.1 | - | 2 | 242 |

More Information
Other Species Targeted

