siRNA Details
Gene Symbol: Slc25a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002283.1 | - | 3 | 801 | ||
AK003467.1 | - | 3 | 805 | |||
AK012750.1 | - | 3 | 807 | |||
AK012751.1 | - | 3 | 803 | |||
AK017881.1 | - | 3 | 800 | |||
AK088730.1 | - | 3 | 802 | |||
AK215039.1 | - | 1 | 37 | |||
BC004570.1 | - | 3 | 785 | 3586336 | ||
BC086756.1 | - | 3 | 784 | 6818521 | ||
U10404.1 | - | 3 | 725 | |||
U27316.1 | - | 3 | 810 | |||
X70847.1 | - | 3 | 812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
