siRNA Details
Gene Symbol: Aoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009675.2 | NP_033805.1 | 1 | 1611 | ||
GenBank mRNA | AF054831.1 | - | 1 | 1626 | ||
AF115411.1 | - | 1 | 1442 | |||
BC080857.1 | - | 1 | 1450 | 6854465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
