siRNA Details
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243043.1 | NP_001229972.1 | 16 | 2612 | ||
NM_001243044.1 | NP_001229973.1 | 15 | 2394 | |||
NM_007454.3 | NP_031480.2 | 15 | 2394 | |||
GenBank mRNA | AA003413.1 | - | 1 | 30 | ||
AK034140.1 | - | 16 | 2615 | |||
AK155892.1 | - | 15 | 2314 | |||
AK159273.1 | - | 15 | 2285 | |||
AK160042.1 | - | 15 | 2285 | |||
AK165747.1 | - | 15 | 2311 | |||
AK170376.1 | - | 15 | 2305 | |||
BC008513.1 | - | 15 | 2207 | 3601435 | ||
BC066827.1 | - | 6 | 938 | |||
Y07919.1 | - | 15 | 2249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
