siRNA Details
Gene Symbol: Ap1m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007456.4 | NP_031482.1 | 5 | 570 | ||
GenBank mRNA | AK148173.1 | - | 5 | 585 | ||
AK192043.1 | - | 2 | 133 | |||
BC003823.1 | - | 5 | 544 | 3491338 | ||
M62419.1 | - | 5 | 572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
