siRNA Details
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 3 | 280 | ||
AK002565.1 | - | 3 | 306 | |||
AK137489.1 | - | 3 | 301 | |||
AK152100.1 | - | 3 | 312 | |||
AK194047.1 | - | 1 | 62 | |||
BC012656.1 | - | 3 | 291 | 4187607 | ||
BC108327.1 | - | 3 | 299 | 30713247 | ||
BC131985.1 | - | 3 | 281 | 40130262 | ||
BC145234.1 | - | 3 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
