siRNA Details
Gene Symbol: Birc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012273.1 | NP_001012273.1 | 3 | 424 | ||
NM_009689.2 | NP_033819.1 | 3 | 424 | |||
GenBank mRNA | AB013819.1 | - | 3 | 418 | ||
AF077349.1 | - | 3 | 325 | |||
AK142768.1 | - | 3 | 340 | |||
BC004702.1 | - | 3 | 342 | 3499203 | ||
CN724527.1 | - | 3 | 424 | |||
EU366955.1 | - | 3 | 310 |
