siRNA Details
Gene Symbol: Birc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012273.1 | NP_001012273.1 | 3 | 404 | ||
NM_009689.2 | NP_033819.1 | 3 | 404 | |||
GenBank mRNA | AB013819.1 | - | 3 | 398 | ||
AF077349.1 | - | 3 | 305 | |||
AK142768.1 | - | 3 | 320 | |||
BC004702.1 | - | 3 | 322 | 3499203 | ||
EU366955.1 | - | 3 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
