siRNA Details
Gene Symbol: Cd5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009690.2 | NP_033820.2 | 4 | 807 | ||
GenBank mRNA | AF011428.1 | - | 4 | 713 | ||
AF018268.1 | - | 4 | 746 | |||
AF018269.1 | - | 4 | 732 | |||
AK159132.1 | - | 4 | 756 | |||
AK159181.1 | - | 4 | 810 | |||
AK159823.1 | - | 4 | 775 | |||
BC006799.1 | - | 4 | 748 | 3152586 | ||
BC094459.1 | - | 4 | 760 | 4161650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
