siRNA Details
Gene Symbol: Apoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013474.2 | NP_038502.2 | 4 | 342 | ||
GenBank mRNA | AA060885.1 | - | 3 | 160 | ||
AK145823.1 | - | 4 | 305 | |||
BC031786.1 | - | 3 | 288 | 4195958 | ||
BU609941.1 | - | 493 | ||||
CB947382.1 | - | 4 | 307 | |||
M79361.1 | - | 4 | 307 | |||
M79362.1 | - | 4 | 307 | |||
S79794.1 | - | 3 | 248 | |||
X04119.1 | - | 3 | 274 | |||
X62772.1 | - | 4 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
