siRNA Details
Gene Symbol: Apoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007468.2 | NP_031494.2 | 3 | 870 | ||
GenBank mRNA | AK004856.1 | - | 3 | 653 | ||
AK161535.1 | - | 3 | 653 | |||
AK168687.1 | - | 3 | 654 | |||
AK169277.1 | - | 3 | 740 | |||
BC010769.1 | - | 3 | 663 | 4240796 | ||
BC050149.1 | - | 3 | 628 | 30013747 | ||
M64248.1 | - | 3 | 652 | |||
M64249.1 | - | 3 | 652 | |||
M64250.1 | - | 3 | 652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
