siRNA Details
Gene Symbol: Apoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110009.2 | NP_001103479.1 | 3 | 184 | ||
NM_007469.5 | NP_031495.1 | 2 | 306 | |||
GenBank mRNA | AK011358.1 | - | 2 | 221 | ||
BC019398.1 | - | 3 | 159 | 5124986 | ||
BC035445.1 | - | 3 | 171 | |||
BC094638.1 | - | 2 | 153 | 875610 | ||
BY087738.1 | - | 3 | 185 | |||
CB947285.1 | - | 2 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
