siRNA Details
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013475.4 | NP_038503.4 | 4 | 414 | ||
GenBank mRNA | AA273732.1 | - | 1 | 29 | ||
AK010926.1 | - | 4 | 402 | |||
BC011037.1 | - | 4 | 430 | |||
BC019811.1 | - | 4 | 402 | 5125770 | ||
BC053338.1 | - | 4 | 390 | 5125963 | ||
D10056.1 | - | 4 | 388 | |||
S70439.1 | - | 4 | 398 | |||
Y11356.1 | - | 4 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
