siRNA Details
Gene Symbol: Arf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130408.1 | NP_001123880.1 | 2 | 421 | ||
NM_007476.3 | NP_031502.1 | 2 | 229 | |||
GenBank mRNA | AK077542.1 | - | 2 | 179 | ||
AK148458.1 | - | 2 | 193 | |||
AK148873.1 | - | 2 | 183 | |||
AK150092.1 | - | 2 | 202 | |||
AK150699.1 | - | 2 | 179 | |||
AK151223.1 | - | 2 | 179 | |||
AK151225.1 | - | 2 | 202 | |||
AK151696.1 | - | 2 | 155 | |||
AK151821.1 | - | 2 | 177 | |||
AK152015.1 | - | 2 | 175 | |||
AK152382.1 | - | 2 | 177 | |||
AK152479.1 | - | 2 | 436 | |||
AK153132.1 | - | 2 | 196 | |||
AK159174.1 | - | 2 | 179 | |||
AK166628.1 | - | 2 | 180 | |||
AK167000.1 | - | 2 | 158 | |||
AK167129.1 | - | 2 | 192 | |||
AK167543.1 | - | 2 | 179 | |||
AK167931.1 | - | 2 | 193 | |||
AK168081.1 | - | 2 | 366 | |||
AK168686.1 | - | 2 | 179 | |||
AK169166.1 | - | 2 | 177 | |||
AK170092.1 | - | 2 | 181 | |||
AK170586.1 | - | 2 | 154 | |||
AK216070.1 | - | 1 | 140 | |||
BC021403.1 | - | 1 | 172 | |||
BC031986.1 | - | 1 | 146 | 4012689 | ||
CN705066.1 | - | 2 | 261 | |||
D87898.1 | - | 2 | 176 |

More Information
Other Species Targeted

Gene Ontology Categories:
