siRNA Details
Gene Symbol: GSTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000846.4 | NP_000837.3 | 7 | 879 | ||
XM_011514532.2 | XP_011512834.1 | 7 | 871 | |||
GenBank mRNA | AK292602.1 | - | 7 | 791 | ||
BC002895.2 | - | 7 | 842 | 3943417 | ||
M16594.1 | - | 7 | 795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
