siRNA Details
Gene Symbol: Rhob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007483.2 | NP_031509.1 | 1 | 469 | ||
GenBank mRNA | AF481943.1 | - | 1 | 116 | ||
AK086873.1 | - | 1 | 494 | |||
AK164518.1 | - | 1 | 494 | |||
AK167917.1 | - | 1 | 494 | |||
BC018275.1 | - | 1 | 469 | 4168565 | ||
BC023334.1 | - | 1 | 225 | |||
BC110992.1 | - | 1 | 480 | 5376565 | ||
CT010379.1 | - | 1 | 116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
