siRNA Details
Gene Symbol: GSTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292602.1 | - | 5 | 396 | ||
AK313601.1 | - | 5 | 396 | |||
BC002895.2 | - | 5 | 447 | 3943417 | ||
BE795593.1 | - | 5 | 427 | |||
BE796134.1 | - | 5 | 428 | |||
M16594.1 | - | 5 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
