siRNA Details
Gene Symbol: Rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009708.1 | NP_033838.1 | 3 | 252 | ||
GenBank mRNA | AF016482.1 | - | 3 | 235 | ||
AK075970.1 | - | 560 | ||||
AK170529.1 | - | 1683 | ||||
BC107022.2 | - | 3 | 251 | |||
BC127034.1 | - | 3 | 246 | |||
BC128475.1 | - | 3 | 251 | |||
BC139330.1 | - | 3 | 325 | 8862352 | ||
BC139331.1 | - | 3 | 325 | 8862353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
