siRNA Details
Gene Symbol: Rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009708.1 | NP_033838.1 | 2, 3 | 198 | ||
GenBank mRNA | AF016482.1 | - | 2, 3 | 181 | ||
AK075970.1 | - | 506 | ||||
BC107022.2 | - | 2, 3 | 197 | |||
BC127034.1 | - | 2, 3 | 192 | |||
BC128475.1 | - | 2, 3 | 197 | |||
BC139330.1 | - | 2, 3 | 271 | 8862352 | ||
BC139331.1 | - | 2, 3 | 271 | 8862353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
