siRNA Details
Gene Symbol: Arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023142.2 | NP_075631.2 | 8 | 1075 | ||
GenBank mRNA | AF162768.1 | - | 7 | 1000 | ||
AK002274.1 | - | 8 | 1076 | |||
AK005378.1 | - | 8 | 1076 | |||
AK147731.1 | - | 8 | 1043 | |||
AK148145.1 | - | 8 | 1069 | |||
AK151911.1 | - | 8 | 1076 | |||
AK153510.1 | - | 8 | 1076 | |||
AK157098.1 | - | 8 | 1066 | |||
AK159371.1 | - | 8 | 1074 | |||
AK159477.1 | - | 8 | 965 | |||
AK167978.1 | - | 8 | 1005 | |||
AK168774.1 | - | 8 | 1073 | |||
AK170759.1 | - | 8 | 1065 | |||
AK171364.1 | - | 8 | 1037 | |||
AK172124.1 | - | 8 | 1043 | |||
BC003441.1 | - | 8 | 1007 | 2650641 | ||
BC010275.1 | - | 8 | 1017 | 3589768 | ||
BC020404.1 | - | 5 | 603 | |||
BC092051.1 | - | 8 | 1031 | 3668682 |

More Information
Other Species Targeted

Gene Ontology Categories:
