siRNA Details
Gene Symbol: Art1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009710.4 | NP_033840.2 | 3 | 827 | ||
GenBank mRNA | AK003107.1 | - | 3 | 828 | ||
AK009902.1 | - | 828 | ||||
BC063760.1 | - | 3 | 808 | 30137479 | ||
BC132492.1 | - | 3 | 815 | 40130769 | ||
BC132496.1 | - | 3 | 815 | 40130773 | ||
X95842.1 | - | 2 | 789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
