siRNA Details
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009712.3 | NP_033842.3 | 3 | 769 | ||
GenBank mRNA | AK083309.1 | - | 3 | 753 | ||
AK140163.1 | - | 3 | 819 | |||
AK154098.1 | - | 3 | 689 | |||
AK158312.1 | - | 3 | 770 | |||
BC150661.1 | - | 3 | 726 | 9087572 | ||
X92096.1 | - | 2 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
