siRNA Details
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009712.3 | NP_033842.3 | 8 | 1528 | ||
GenBank mRNA | AK140163.1 | - | 8 | 1583 | ||
AK154098.1 | - | 8 | 1448 | |||
AK195674.1 | - | 2 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
