siRNA Details
Gene Symbol: Atp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009724.2 | NP_033854.1 | 2 | 159 | ||
GenBank mRNA | AK008180.1 | - | 2 | 158 | ||
AK008686.1 | - | 2 | 159 | |||
AK009499.1 | - | 2 | 159 | |||
AK009670.1 | - | 2 | 164 | |||
AK009832.1 | - | 2 | 161 | |||
AK009994.1 | - | 2 | 158 | |||
AK010003.1 | - | 2 | 158 | |||
AK010030.1 | - | 2 | 159 | |||
AK010049.1 | - | 2 | 206 | |||
BC120705.1 | - | 2 | 158 | 40129628 | ||
BC125292.1 | - | 2 | 158 | 40129807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
