siRNA Details
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007505.2 | NP_031531.1 | 4 | 650 | ||
GenBank mRNA | AK043976.1 | - | 4 | 418 | ||
AK076572.1 | - | 4 | 418 | |||
AK098155.1 | - | 4 | 428 | |||
AK146797.1 | - | 4 | 421 | |||
AK150426.1 | - | 4 | 421 | |||
AK150843.1 | - | 4 | 418 | |||
AK151004.1 | - | 4 | 418 | |||
AK151128.1 | - | 4 | 427 | |||
AK151224.1 | - | 4 | 425 | |||
AK151920.1 | - | 4 | 422 | |||
AK152054.1 | - | 4 | 429 | |||
AK152890.1 | - | 4 | 421 | |||
AK153167.1 | - | 4 | 422 | |||
AK153282.1 | - | 4 | 418 | |||
AK159491.1 | - | 4 | 418 | |||
AK159540.1 | - | 4 | 418 | |||
AK159758.1 | - | 4 | 421 | |||
AK160043.1 | - | 4 | 428 | |||
AK164110.1 | - | 4 | 421 | |||
AK164193.1 | - | 2 | 124 | |||
AK166709.1 | - | 4 | 421 | |||
AK166812.1 | - | 4 | 434 | |||
AK167159.1 | - | 4 | 418 | |||
AK167863.1 | - | 4 | 398 | |||
AK168198.1 | - | 4 | 428 | |||
AK168369.1 | - | 4 | 418 | |||
AK168617.1 | - | 4 | 418 | |||
AK168879.1 | - | 4 | 418 | |||
AK168890.1 | - | 4 | 418 | |||
AK168932.1 | - | 4 | 418 | |||
AK169080.1 | - | 4 | 421 | |||
AK169084.1 | - | 4 | 421 | |||
AK169105.1 | - | 4 | 418 | |||
AK169142.1 | - | 4 | 463 | |||
AK169300.1 | - | 4 | 418 | |||
AK169308.1 | - | 4 | 423 | |||
AK169414.1 | - | 4 | 421 | |||
AK192613.1 | - | 2 | 177 | |||
AK196974.1 | - | 2 | 177 | |||
AK209959.1 | - | 2 | 177 | |||
AK218124.1 | - | 4 | 347 | |||
BC014854.1 | - | 4 | 405 | 4500893 | ||
BC020417.1 | - | 4 | 405 | |||
BC055892.1 | - | 4 | 855 | |||
L01062.1 | - | 4 | 405 |

More Information
Other Species Targeted

Gene Ontology Categories:
